Learn More
Medchemexpress LLC Izuforant (JW1601) | 1429374-83-3 | 98.9% | 334.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1472765MG
Encompass_Preferred
Izuforant (JW1601) is an orally active histamine H4 receptor (H4R) antagonist with an IC50 of 36 nM against human H4R. It also shows binding affinity to the human serotonin 3 receptor (h5-HT3R) with an IC50 of 9.1 μM. This compound exhibits strong anti-pruritic and anti-inflammatory efficacies and is suitable for research applications.
  • Orally active histamine H4 receptor antagonist
  • Binds to human serotonin 3 receptor
  • Exhibits strong anti-pruritic and anti-inflammatory efficacies
  • Shows metabolic stability in mouse and human
  • Decreases histamine- and substance P-induced itching
  • Inhibits oxazolone-induced atopic dermatitis
Catalog No. 50-003-60643