Learn More
Medchemexpress LLC J-2156 | 848647-56-3 | 99.88% | 468.57 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111615
Encompass_Preferred
J-2156 is a highly potent, selective somatostatin receptor type 4 (SST4 receptor) agonist with IC50s of 0.05 nM and 0.07 nM for human and rat SST4 receptors, respectively. It is used for the relief of mechanical allodynia and mechanical hyperalgesia in the ipsilateral hindpaws in rats.
  • Highly potent, selective somatostatin receptor type 4 (SST4 receptor) agonist.
  • IC50s of 0.05 nM (human SST4) and 0.07 nM (rat SST4).
  • Exhibits over 400-fold subtype-selectivity against other somatostatin receptors.
  • Has anti-allodynic effect on ipsilateral and contralateral in BCIBP-rats.
Catalog No. 50-004-39696