Learn More
Medchemexpress LLC J-2156 | 848647-56-3 | 99.9% | 468.57 | C24H28N4O4S | 5 MG

Supplier: Medchemexpress LLC HY1116155MG
J-2156 is a potent, selective somatostatin receptor 4 (SST4) agonist used in research to probe SST4-mediated signaling and inflammatory pain models. It displays subnanomolar activity against human and rat SST4 receptors and is supplied as a high-purity research compound for in vitro and in vivo pharmacology studies.
- Potent, selective SST4 agonist with subnanomolar activity.
- High purity suitable for research use.
- Molecular weight 468.57; formula C24H28N4O4S.
- Soluble in DMSO at 200 mg/mL (requires ultrasonic).
- Powder storage: -80°C (2 years) or -20°C (1 year); in solution: -80°C (6 months) or -20°C (1 month).
- Multiple package sizes available to suit experimental needs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.