Learn More
Chemglass Life Sciences J-KEM Male standard/female mini Thermocouple K
SDP
Supplier:  Chemglass Life Sciences CG321210
Encompass_Preferred
J-KEM Male standard/female mini Thermocouple K. Type K input: -50 to 1200°C
Catalog No. CG321210