Learn More
Medchemexpress LLC J147 | 1807913-16-1 | 99.5% | 350.34 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY13779500MG
Encompass_Preferred
J147 is an exceptionally potent, orally active, neuroprotective agent designed for cognitive enhancement. It readily passes the blood-brain barrier (BBB) and exhibits inhibitory activity against monoamine oxidase B (MAO B) and the dopamine transporter. This compound holds potential for research into Alzheimer's disease (AD).
  • Exceptionally potent and orally active
  • Neuroprotective agent for cognitive enhancement
  • Readily passes the blood-brain barrier
  • Inhibits monoamine oxidase B
  • Inhibits dopamine transporter
  • Potential for Alzheimer's disease research
  • Promotes HT22 and primary cell survival
  • Enhances memory and dendritic spine number
Catalog No. 50-003-50663