Learn More
Beckman Coulter JA-20 Fixed-Angle Aluminum Rotor, 8 x 50 mL, 20,000 rpm, 48,400 x g
Supplier:  Beckman Coulter 334831
Encompass
JA-20 Fixed-Angle Rotor, Aluminum, and Biosafety Lid. Major applications: Harvesting bacteria and cell membranes, processing tissue homogenates, separating cell particulates. For use in Avanti J Series centrifuges.
Catalog No. NC1623694
Only null left
Add to Cart
Request a Quote