Learn More
Beckman Coulter JA18 1 FIXED ANGLE 24 X 1 8ML
Supplier:  Beckman Coulter 347824
Encompass
JA18.1 fixed angle, 24 x 1.8mL rotor,18,000 rpm, 42,100 x g
Catalog No. NC2830051
Add to Cart
Request a Quote