Learn More
Medchemexpress LLC JAB-3068 hydrochloride (synonyms: SHP2-IN-6 hydrochloride) | 2169223-49-6 | 98.9% | 1 ML

Supplier: Medchemexpress LLC HY131132A10MM/1ML
JAB-3068 hydrochloride (SHP2-IN-6) is a potent SHP2 inhibitor with an IC50 of 25.8 nM. It inhibits the proliferation of KYSE-520 cells with an IC50 of 2.17 μM in vitro. This compound is derived from patent WO2017211303A1, compound 7.
- Potent SHP2 inhibitor with an IC50 of 25.8 nM.
- Inhibits KYSE-520 cell proliferation with an IC50 of 2.17 μM.
- Molecular weight: 513.00.
- Chemical formula: C22H27ClF2N6O2S.
- Appears as a light yellow to yellow solid.
- Solubility in DMSO is 115 mg/mL (224.17 mM).
- For in solvent storage: -80°C for 6 months or -20°C for 1 month.
- Store at -20°C in sealed conditions, away from moisture and light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.