Learn More
Medchemexpress LLC Jab-3068 (SHP2-IN-6) | 2169223-48-5 | 99.4% | C22H26F2N6O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13113210MG
Encompass_Preferred
JAB-3068 (SHP2-IN-6) is an orally bioavailable, small-molecule allosteric inhibitor of SHP2 (PTPN11) developed as an oncology drug candidate. It has reported potent SHP2 inhibitory activity (IC50 25.8 nM), a molecular formula of C22H26F2N6O2S, and is supplied as a high-purity, research-grade compound used in preclinical and clinical investigations.
  • Potent SHP2 inhibition with reported IC50 of 25.8 nM.
  • Orally bioavailable small-molecule profile suitable for in vivo studies.
  • Chemical formula C22H26F2N6O2S and molecular weight ~476.5 g/mol.
  • High reported purity suitable for research applications.
  • Used in preclinical and clinical investigations of oncologic indications.
Catalog No. 50-002-60630