Learn More
Bioworld JACALINARTOCARPUS INTEGRI
Supplier:  Bioworld 201200012
Encompass
JACALIN (ARTOCARPUS INTEGRIFOLIA)- SEPAROPORE 4B-CL: Jacalin, D-galactose-specific lectin from jackfruit, interacts with human IgA and one or two other serum proteins. Jacalin coupled Separpore is used for the purification of glycoprote...
Catalog No. 50-197-516