Learn More
Medchemexpress LLC HY-N0831 1mg Medchemexpress, Jaceosidin CAS:18085-97-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08311MG
Encompass_Preferred
Medchemexpress, HY-N0831 1mg Jaceosidin CAS:18085-97-7 Jaceosidin is a flavonoid isolated from Artemisia vestita, induces apoptosis in cancer cells, activates Bax and down-regulates Mcl-1 and c-FLIP expression [1] . Jaceosidin exhibits anti-cancer [2] , anti-inflammatory activities, decreases leves of inflammatory markers, and suppresses COX-2 expression and NF-κB activation [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3736