Learn More
Chemglass Life Sciences Jacketed Beaker, 100 x 50mm ID, 135 x 70mm OD
SDP
Supplier:  Chemglass Life Sciences CG110710
Encompass_Preferred
Jacketed Beaker, 100 x 50mm ID, 135 x 70mm OD. Similar to CG-1103, but with a larger opening at the top of the jacket and a shorter overall height. Width to Height ratio on all sizes is 2:1. Beakers also have a FLAT Bottom on both the inner and outer vessel. Hose Connections have an O.D. of 10mm at the largest serration.
Catalog No. 50-872-932