Learn More
Medchemexpress LLC Jak-in-1 | 1334673-53-8 | 98.4% | 380.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13827100MG
Encompass_Preferred
JAK-IN-1 is a JAK1/2/3 inhibitor with IC50s of 0.26, 0.8, and 3.2 nM, respectively. It exhibits improved selectivity for JAK3 over JAK1. This compound inhibits the proliferation of human CD4 and CD8 T cells in a dose-dependent manner upon stimulation and is active in both mechanistic and functional cell-based assays using T-cells. In vivo, it is JAK3 selective, potently inhibits IL-2 stimulated plasma concentrations, and prevents IL-2-driven STAT5 phosphorylation.
  • Inhibits proliferation of human CD4 and CD8 T cells in a dose-dependent manner.
  • Active in both mechanistic and functional cell-based assays using T-cells.
  • Exhibits JAK3 selectivity in vivo.
  • Potently inhibits IL-2 stimulated plasma concentrations.
  • Prevents IL-2-driven STAT5 phosphorylation in a dose- and concentration-dependent manner.
Catalog No. 50-003-50987