Learn More
Medchemexpress LLC JAK1-IN-8 | 1973485-18-5 | MFCD30530428 | 95.0% | C22H23FN4O3S | 10MG

Supplier: Medchemexpress LLC HY13942310MG
JAK1-IN-8 is a small-molecule research compound reported as a potent Janus kinase 1 (JAK1) inhibitor (IC50 < 500 nM). It corresponds to compound 28 described in patent WO2016119700A1 and is provided as a white to off-white solid for in vitro research applications. Chemical identifiers include CAS 1973485-18-5 and formula C22H23FN4O3S.
- Potent JAK1 inhibition (IC50 < 500 nM).
- Suitable for JAK/STAT pathway and target validation studies.
- Provided as a white to off-white solid for laboratory use.
- Purity typically 95.0% by HPLC.
- Molecular weight 442.51 g/mol; formula C22H23FN4O3S.
- Available in multiple small-scale quantities for research.
- For research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.