Learn More
Medchemexpress LLC JAK3-IN-1 | 1805787-93-2 | 99.3% | 508.02 g/mol | C26H30ClN7O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1954410MG
Encompass_Preferred
JAK3-IN-1 is a potent, selective, orally active inhibitor of Janus kinase 3 used as a research tool to probe JAK/STAT signaling. It exhibits low-nanomolar potency against JAK3 with markedly reduced activity against related kinases, and is supplied as a high-purity solid appropriate for biochemical and cellular studies.
  • Potent JAK3 inhibition (IC50 ~4.8 nM)
  • High selectivity versus JAK1 and JAK2
  • Suitable for biochemical and cellular assays
  • High purity for research applications (~99.3%)
  • Available in small-scale quantities for screening and profiling
Catalog No. 50-002-98302