Learn More
Sino Biological Peptide Substrates: JAK3tide
Supplier:  Sino Biological J03581000
Encompass_Preferred
The JAK3tide synthetic peptide [GGEEEEYFELVKKKK] is routinely evaluated as a substrate for JAK3 and JAK2.
Catalog No. 50-256-8919