Learn More
Med Vet International Miltex Vantage Jansen Bayonet Dressing Forceps, 6.25in
Supplier:  Med Vet International MILV96197
Encompass_Preferred
The Miltex Vantage V96-197 Jansen bayonet dressing forceps are made from high quality Japanese stainless steel and measure 6.25" in length. They have a non-locking thumb handle and are bayonet-shaped. The tips are straight and serrated.
Catalog No. 50-255-9454