Learn More
ABclonal Technology JARID1B Rabbit pAb
SDP
Supplier:  ABclonal Technology A14104500UL
Encompass_Preferred
This gene encodes a lysine-specific histone demethylase that belongs to the jumonji/ARID domain-containing family of histone demethylases. The encoded protein is capable of demethylating tri-, di- and monomethylated lysine 4 of histone H3. This protein plays a role in the transcriptional repression or certain tumor suppressor genes and is upregulated in certain cancer cells. This protein may also play a role in genome stability and DNA repair. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-08373