Learn More
Medchemexpress LLC HY-115521 1mg Medchemexpress, Jarin-1 CAS:1212704-51-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1155211MG
Encompass_Preferred
Medchemexpress, HY-115521 1mg Jarin-1 CAS:1212704-51-2 Jarin-1 is a jasmonic acid-amido synthetase (JAR1) inhibitor with an IC50 of 3.8 μM. Jarin-1 specific inhibits bioactive JA (jasmonoyl-isoleucine, JA-Ile) biosynthesis in Arabidopsis and other plants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9390