Learn More
TARGETMOL CHEMICALS INC JARIN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T156085MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Jarin-1 is a jasmonic acid-amido synthetase (JAR1) inhibitor (IC50 3.8 uM). Jarin-1 specific suppresses bioactive JA biosynthesis in Arabidopsis and other plants. purity: 99%
Catalog No. 50-258-1218