Learn More
Optimize Technologies OPTI-MAX™ Cartridge Check Valve Conversion Kit, Jasco BIP-1
SDP
Supplier:  Optimize Technologies 425602150
Encompass_Preferred
Make the switch to OPTI-MAX™ for less than ever before with OPTI-MAX™ Conversion Kits. Each kit includes all the OPTI-MAX™ check valve housings and cartridges you need to convert your HPLC instrument. By default, each kit includes our recommended OPTI-MAX™ cartridge, stainless steel/ceramic OPTI-MAX™ (PEEK cartridges are supplied for biocompatible systems).
Catalog No. 50-200-6719