Learn More
Cayman Chemical JasmonIc AcId methyl ester 1g
SDP
Supplier:  Cayman Chemical 90000591 G
Encompass_Preferred
Induces the synthesis of proteinase inhibitors in plant leaves; in cancer cells, suppresses proliferation and induces apoptosis; inhibits hexokinase that is bound to mitochondria; methyl jasmonate derivatives also have potential as anti-inflammatory agents.
Catalog No. 50-289-6506