Learn More
Cayman Chemical Jasmonic AcidIsoleucine
SDP
Supplier:  Cayman Chemical 107401 MG
Encompass_Preferred
A plant hormone formed by the conjugation of jasmonic acid and isoleucine; signals through COI1-JAZ to direct the degradation of JAZ proteins
Catalog No. 50-288-8259