Learn More
AdipoGen Jasplakinolide (high purity) (50 µg)
SDP
Supplier:  AdipoGen AGCN20037C050
Encompass_Preferred
Jasplakinolide (high purity) (50 µg), AdipoGen Life Sciences. Chemical. CAS: 102396-24-7. Formula: C36H45BrN4O6. MW: 709.7. Isolated from Jaspis splendens. Cell permeable, non-fluorescent F-actin probe. Potent inducer of actin polymerization and stabilization. Competes with phallotoxins for actin binding. Antifungal and antiparasitic compound. Antiproliferative and anticancer compound. Apoptosis enhancer/inducer. Tool used for autophagy/phagocytosis research.
Catalog No. 50-148-9963