Learn More
Enzo Life Sciences Jasplakinolide, (50µg), CAS Number: 102396-24-7
SDP
Supplier:  Enzo Life Sciences ALX350275C050
Encompass_Preferred
F-actin probe. Cell permeable F-actin probe. Induces actin polymerization. Completely inhibits the binding of phalloidin to F-actin. Antifungal and anti-proliferative agent. Alternative Name: Jaspamide. Formula: C36H45BrN4O6. MW: 709.7. CAS Number: 102396-24-7. Purity: ≥97% (HPLC). Appearance: Off-white to yellow solid. Solubility: Soluble in 100% ethanol, 83% methanol or DMSO. Long Term Storage: -20°C
Catalog No. 50-103-1744