Learn More
Medchemexpress LLC Jatrorrhizine chloride | 6681-15-8 | 373.83 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN07401MG
Encompass_Preferred
Jatrorrhizine chloride is an alkaloid isolated from Coptis chinensis, exhibiting neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. This product is for research use only and not sold to patients.
  • Isolated from Coptis chinensis
  • Exhibits neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities
  • Potent and orally active inhibitor of AChE (IC50=872 nM) with over 115-fold selectivity for BuChE
  • Reduces uptake of serotonin (5-HT) and norepinephrine (NE) via inhibition of uptake-2 transporters
Catalog No. 50-004-15784