Learn More
Medchemexpress LLC Jatrorrhizine (chloride) | 6681-15-8 | 373.83 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN074010MM/1ML
Encompass_Preferred
Jatrorrhizine chloride is an alkaloid isolated from Coptis chinensis that possesses neuroprotective, antimicrobial, antiplasmodial, and antioxidant activities. It acts as a potent and orally active inhibitor of AChE (IC50=872 nM), demonstrating over 115-fold selectivity for BuChE. Additionally, it reduces the uptake of serotonin (5-HT) and norepinephrine (NE) by inhibiting uptake-2 transporters.
  • Neuroprotective activity
  • Antimicrobial activity
  • Antiplasmodial activity
  • Antioxidant activity
  • Potent and orally active inhibitor of AChE
  • Over 115-fold selectivity for BuChE
  • Reduces uptake of serotonin (5-HT) and norepinephrine (NE)
  • Inhibits PMAT-mediated MPP+ uptake
  • Inhibits 5-HT and NE uptake in synaptosomes
Catalog No. 50-004-15806