Learn More
Medchemexpress LLC Jb061 | C19H17NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15220510MM/1ML
Encompass_Preferred
JB061 is a nonmuscle myosin inhibitor known for its specific inhibition of various myosin types and its cytotoxic effects on COS-7 cells. It effectively inhibits cytokinesis in Cos-7 cells.
  • Nonmuscle myosin inhibitor
  • Inhibits cytokinesis in Cos-7 cells
  • High purity (99.27%)
  • Solid appearance, white to off-white color
  • Stable storage conditions for powder and in solvent
Catalog No. 50-003-73795