Learn More
ATCC JB6 Cl 41-5a Mouse Epidermal Cell Line (ATCC CRL-2010) - Tumor Promotion Assay Model
Supplier:  ATCC JB6 CL 415A CRL2010
Encompass
JB6 Cl 41-5a (ATCC CRL-2010) is a mouse epidermal cell line established from neonatal BALB/c epidermal cells. It is adherent and sensitive to promotion of transformation by phorbol esters, making it useful for in vitro studies of tumor promotion and related molecular pathways.
  • Derived from neonatal BALB/c mouse epidermal cells.
  • Adherent growth properties.
  • Sensitive to promotion of transformation by phorbol esters.
  • Used in tumor promotion and transformation assays.
  • Suitable for in vitro molecular studies of promotion pathways.
Catalog No. NC3720597