Learn More
Medchemexpress LLC JBI-589 | 2308504-22-3 | C29H28FN5O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1534501MG
Encompass_Preferred
JBI-589 is a non-covalent, orally bioavailable, and isoform-selective inhibitor of PAD4. It reduces CXCR2 expression and blocks neutrophil chemotaxis, and has been shown to reduce primary tumor and metastases. This compound enhances the anti-tumor effect of checkpoint inhibitors, making it suitable for cancer research.
  • Non-covalent and orally bioavailable
  • Isoform-selective inhibitor of PAD4
  • Reduces CXCR2 expression and blocks neutrophil chemotaxis
  • Inhibits primary tumor growth and metastases
  • Enhances anti-tumor effect of checkpoint inhibitors
Catalog No. 50-003-72775