Learn More
Medchemexpress LLC Jbj-09-063 hydrochloride | 99.37% | 593.11 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY147183B10MM/1ML
Encompass_Preferred
JBJ-09-063 hydrochloride is a mutant-selective allosteric EGFR inhibitor. It effectively reduces EGFR, Akt, and ERK1/2 phosphorylation and is effective across EGFR tyrosine kinase inhibitor (TKI)-sensitive and resistant models. This compound can be used for researching EGFR-mutant lung cancer.
  • Mutant-selective allosteric EGFR inhibitor
  • Effectively reduces EGFR, Akt, and ERK1/2 phosphorylation
  • Effective across TKI-sensitive and resistant models
  • Suitable for researching EGFR-mutant lung cancer
Catalog No. 50-003-62488