Learn More
Medchemexpress LLC JBJ-09-063 TFA, 10mg | Molecular Weight- 670.67 | Formula- C33H30F4N4O5S
SDP
Supplier:  Medchemexpress LLC HY147183A10MG
Encompass_Preferred
JBJ-09-063 TFA is a mutant-selective allosteric EGFR inhibitor with IC50s of 0.147 nM, 0.063 nM, 0.083 nM and 0.396 nM for EGFR L858R, EGFR L858R/T790M, EGFR L858R/T790M/C797S and EGFRLT/L747S. JBJ-09-063 TFA effectively reduces EGFR, Akt and ERK1/2 phosphorylation. JBJ-09-063 TFA is effective across EGFR tyrosine kinase inhibitor (TKI)-sensitive and resistant models. JBJ-09-063 TFA can be used for researching EGFR-mutant lung cancer.
Catalog No. NC2951155