Learn More
AdipoGen JC-1 ,Chemical. CAS: 3520-43-2 and 47729-63-5
SDP
Supplier:  AdipoGen AGCR13568M001
Encompass_Preferred
JC-1 , AdipoGen Life Sciences. Chemical. CAS: 3520-43-2 and 47729-63-5. Formula: C25H27Cl4IN4. MW: 652.2. The membrane-permeant dual-emission potential-sensitive JC-1 dye is widely used in apoptosis studies to monitor mitochondrial health by flow cytometry, fluorescence microscopy and in microplate-based fluorescent assays. JC-1 dye can be used as an indicator of mitochondrial membrane potential in a variety of cell types, including myocytes and neurons, as well as in intact tissues and isolated mitochondria. JC-1 accumulates in mitochondria, selectively generating an orange J-aggregate emission profile (590 nm) in healthy cells. After cell injury, as membrane potential decreases, JC-1 monomers are generated, resulting in a shift to green emission (529 nm).
Catalog No. 50-464-759