Learn More
Apexbio Technology LLC JC-1(Synonyms: 5,5',6,6'-Tetrachloro-1,1',3,3'-tetraethylbenzimidazolylcarbocyanine iodide), 200mg, CAS: 3520-43-2.
SDP
Supplier:  Apexbio Technology LLC A3516200
Encompass_Preferred
JC-1 (CAS 3520-43-2) is a fluorescent probe commonly utilized to evaluate mitochondrial membrane potential thereby serving as a functional indicator of mitochondrial activity Within cells JC-1 selectively accumulates in mitochondria in a membrane-potential-dependent manner shifting fluorescence emission from green to red as it forms aggregates at higher mitochondrial potentials This fluorescence shift allows researchers to monitor mitochondrial membrane integrity and assess cellular metabolic states JC-1 is widely applied in studies of apoptosis mitochondrial dysfunction and cellular bioenergetics
Catalog No. 50-000-60776