Learn More
Selleck Chemical LLC JC-1 S9784-5MG
SDP
Supplier:  Selleck Chemical LLC S97845MG
Encompass_Preferred
JC-1 (CBIC2 NK 1420) a fluorescent lipophilic carbocyanine dye is a mitochondrial potential ( (m)) marker JC-1 fluorescence is usually excited by the 488 nm laser wavelength common in flow cytometers
Catalog No. 50-000-24801