Learn More
TARGETMOL CHEMICALS INC JC2-11 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7757910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. JC2-11 is an inhibitory compound for inflammation.JC2-11 inhibits the recruitment domain-containing proteins NLRC 4, atypical in melanoma 2 (AIM 2) and atypical (NC) inflammatory vesicles by disrupting reactive oxygen species (ROS) production and caspase-1 activity.JC2-11 reduces caspase-1 (p20) secretion, gasdermin D (GSDMD) cleavage, IL-1beta and lactate dehydrogenase (LDH) activity in inflammatory vesicles. JC2-11 reduced caspase-1 (p20) secretion, gasdermin D (GSDMD) cleavage, IL-1beta and lactate dehydrogenase (LDH) release from inflammatory vesicles. Purity 98.6%
Catalog No. 50-287-9298