Learn More
Medchemexpress LLC JCN037 (JGK037) | 2305154-31-6 | 99.1% | 376.18 | 1 ML

Supplier: Medchemexpress LLC HY13643010MM/1ML
JCN037 (JGK037) is a non-covalent and BBB-penetrant EGFR tyrosine kinase inhibitor, with IC50 values of 2.49 nM, 3.95 nM, and 4.48 nM for EGFR, p-wtEGFR, and pEGFRvⅢ, respectively. It exhibits GI50 values of 329 nM in HK301 cells and 1116 nM in GBM39 cells. In vivo, it downregulates pEGFRvⅢ, p Akt, p-ERK, and p-S6 protein levels and provides a significant survival benefit, increasing median survival by 47% from 37.5 days to 55 days.
- Non-covalent EGFR tyrosine kinase inhibitor.
- BBB-penetrant.
- Inhibits EGFR (IC50: 2.49 nM).
- Effective in HK301 (GI50: 329 nM) and GBM39 (GI50: 1116 nM) cells.
- Downregulates key protein phosphorylation.
- Increases median survival by 47% in vivo.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.