Learn More
Molecular Dimensions JCSG-PLUS HT-96
Supplier: Molecular Dimensions MD140
Protein Crystallography. JCSG-plus screen HT-96 (96 x 1.1mL) is the screen of choice for initial screening experiments.The most complete sparse matrix screen available today. Screen classic PEG and salt conditions more efficiently. Modern precipitants. Range of salts.Broad pH range.