Learn More
Jeio Tech Inc Jeio Tech™ IB4-10AV, 114L Advanced Air Jacket Incubator (120V, 60Hz), PID Temperature Control, Uniform Heating, Temperature Ramping, Data Logging (RS-232/USB)
SDP
Supplier:  Jeio Tech Inc AAH21773U
Encompass_Preferred
Jeio Tech IB4-10AV Air-Jacket Advanced Incubator delivers precise and uniform temperature control using a high-performance air-jacket heating system, ideal for sensitive biological and clinical applications. The air-jacket design ensures gentle and stable temperature distribution, reducing the risk of thermal fluctuations and protecting delicate cultures. Equipped with an intuitive digital PID controller, it provides excellent stability (±0.15°C fluctuation) and uniformity (±0.46°C variation at 37°C), while the advanced model includes features such as fan speed control, programmable temperature ramping, real-time graphical monitoring, and RS-232/USB communication ports for data logging and remote access. With a durable corrosion-resistant inner chamber (W 19.1" × D 13.78" × H 25.79".), adjustable shelving, and built-in safety protections, the IB4-A is the optimal choice for reliable incubation in microbiology, biotechnology, and pharmaceutical laboratories.
Catalog No. NC3965866