Learn More
Jeio Tech Inc Jeio Tech™ MS-51M Multi Position Magnetic Stirrer (5 Rows x 1 Column, 100-240V, 50/60Hz), Feedback Speed Control, BLDC Motor, Strong Magnetic Coupling, Timer, Spill-Proof Design, Quiet Mixing
SDP
Supplier:  Jeio Tech Inc AAH332415U
Encompass_Preferred
Jeio Tech MS-51M Multi-Position Magnetic Stirrer, High-Throughput Mixing with Precise Speed Control delivers efficient and flexible performance for demanding lab workflows. Featuring feedback control and a wide speed range (30-2000 rpm), it ensures accurate, consistent stirring across all positions. The durable BLDC motor provides quiet, maintenance-free operation, while strong permanent magnets enable stable magnetic coupling. Flexible control options allow independent point or column operation to suit various applications. The smooth start/stop function minimizes splashing for safer handling, and the spill-proof design protects internal components. A built-in timer supports programmable mixing, and an optional silicone pad enhances vessel stability. With optimized spacing (90mm), the MS-51M is ideal for reliable, high-throughput laboratory mixing.
Catalog No. NC3965956