Learn More
MSC Jergens Center-Pull Hoist Ring, 2500 lb Capacity, 1/2-13 Thread, Alloy Steel
Supplier:  MSC 94455201
Encompass
This center-pull hoist ring is manufactured in the USA from certified alloy steel, ensuring robust and reliable quality for heavy-duty lifting applications. It features a design that meets rigorous safety standards for secure lifting.
  • Material is alloy steel with a black oxide finish.
  • Washer material is zinc plated with gold color.
  • Critical components undergo magnetic particle inspection.
  • Conforms to ASME B30.26 and MIL-STD-209K standards.
  • Rated with a load capacity of 2500 lb and a 1/2-13 thread.
  • Thread length measures 1 1/4 in.
  • Installation requires a torque of 28 ft. lbs.
Catalog No. 50-005-30461