Learn More
Medchemexpress LLC Jervine (11-Ketocyclopamine) | 469-59-0 | 425.60 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0836100MG
Encompass_Preferred
Jervine (11-Ketocyclopamine) is a natural teratogenic steroidal alkaloid derived from *Veratrum nigrum*. It acts as a potent Hedgehog (Hh) inhibitor and exhibits anti-inflammatory and antioxidant properties.
  • Potent Hedgehog (Hh) inhibitor
  • Natural steroidal alkaloid
  • Possesses anti-inflammatory properties
  • Exhibits antioxidant properties
  • Inhibits Akt phosphorylation and NF-κB activation
  • Decreases COX-2 overexpression
  • Induces apoptosis in vitro
  • Demonstrates anti-inflammatory effects in vivo
Catalog No. 50-004-16796