Learn More
Electron Microscopy Sciences Jewelers Saw Blade Size 2/0
SDP
Supplier:  Electron Microscopy Sciences 7202002
Encompass_Preferred
Swiss-made jewlers saw with a fully adjustable frame that permits the tightening of the blade as well as using broken blades. Each blade has an overall length of 5¼" (133 mm).
Thickness: 0.25 mm
Width: 0.53 mm
Teeth: 21 teeth per cm
Catalog No. 50-949-098