Learn More
Electron Microscopy Sciences DISCONTINUED
SDP
Supplier:  Electron Microscopy Sciences 72010
Encompass_Preferred
Swiss-made jewlers saw with a fully adjustable frame that permits the tightening of the blade as well as using broken blades. Each blade has an overall length of 5" (133 mm). Comes with 12 blades (size 3/0).
Catalog No. 50-949-231