Learn More
Selleck Chemical LLC JG98 2mg
SDP
Supplier:  Selleck Chemical LLC S67212MG
Encompass_Preferred
JG-98 is an allosteric inhibitor ofHsp70 that binds tightly to a deep pocket that is conserved in members of the Hsp70 family. JG-98 induces classicalapoptosis features, including morphological changes consistent with programmed cell death and positive annexin staining. JG-98 exhibits anticancer activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3214