Learn More
TARGETMOL CHEMICALS INC JH-RE-06 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T156115MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JH-RE-06 disrupts mutagenic translesion synthesis (TLS) by preventing the recruitment of mutagenic POL(zeta). JH-RE-06 is an effective REV1-REV7 interface inhibitor (IC50=0.78 uM; Kd=0.42 uM) which targets REV1 that interacts with the REV7 subunit of POL(zeta). JH-RE-06 also improves chemotherapy. purity: 99%
Catalog No. 50-258-1216