Learn More
TARGETMOL CHEMICALS INC JH-VIII-157-02 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T156121MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JH-VIII-157-02 is an inhibitor of ALK and inhibits echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) with IC50s of 2 nM for EML4-ALK G1202R, EML4-ALKwt, EML4-ALK C1156Y, EML4-ALK F1174L, and EML4-ALK F1174L. Purity 98.02%
Catalog No. 50-287-8487