Learn More
Medchemexpress LLC Jh-x-119-01 | 2227368-54-7 | MFCD34593626 | 99.0% | 452.46 g/mol | C25H20N6O3 | 5 MG

Supplier: Medchemexpress LLC HY103017A5MG
Small-molecule IRAK1 inhibitor intended for laboratory research use, suitable for in vitro and in vivo studies. Supplied as a solid with high purity and characterized by molecular formula C25H20N6O3 and molecular weight 452.46 g/mol.
- Selective IRAK1 inhibitor for research applications.
- High purity (99.03%) suitable for biological assays.
- Molecular formula C25H20N6O3, molecular weight 452.46 g/mol.
- Appearance: solid, light yellow to brown.
- Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
- Available in small milligram quantities, including 5 mg vials.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.