Learn More
Medchemexpress LLC JH-X-119-01 hydrochloride | 2591344-30-6 | 99.1% | 488.93 g/mol | C25H21ClN6O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10301750MG
Encompass_Preferred
JH-X-119-01 hydrochloride is a research-grade small-molecule inhibitor of interleukin-1 receptor-associated kinase 1 (IRAK1). It is supplied as a solid with high reported purity and has demonstrated activity in preclinical LPS-induced sepsis models. Intended for laboratory research use only.
  • Potent, selective IRAK1 inhibitor.
  • Demonstrated efficacy in LPS-induced sepsis mouse models.
  • High purity suitable for research applications.
  • Solid form, light yellow to yellow in color.
  • Provided in small milligram packaging for preclinical studies.
Catalog No. 50-002-47566