Learn More
Medchemexpress LLC JH-XII-03-02 | 2415900-86-4 | 99.4% | 853.92 g/mol | C43H51N9O10 | 10 MG

Supplier: Medchemexpress LLC HY15515010MG
JH-XII-03-02 is a potent and selective PROTAC degrader of leucine-rich repeat kinase 2 (LRRK2) intended for Parkinson's disease research. It promotes degradation of LRRK2 and inhibits kinase activity, enabling mechanistic studies of LRRK2 function, signal transduction, and disease-relevant cellular phenotypes in vitro.
- Potent, selective LRRK2 PROTAC degrader.
- High purity: 99.41% (reported).
- Molecular weight: 853.92 g/mol.
- Molecular formula: C43H51N9O10.
- CAS number: 2415900-86-4.
- Useful for studies of protein degradation and kinase inhibition.
- Supplied as a 10 mg research quantity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.